Assay Design



Gene Transcripts

Gene Symbol
CLIP3
Entrez Gene ID
25999
Gene Name
CAP-Gly domain containing linker protein 3
Gene Aliases
CLIPR-59, CLIPR59, RSNL1
Chromosome Location
Chr.19: 36014660 - 36032895 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199570.1 NP_001186499.1 4 - 5 875 59
NM_015526.2 NP_056341.1 5 - 6 791 59
GenBank mRNA AJ427922.1 - 5 - 6 769 59
AK056445.1 - 3 - 4 275 59
AK090732.1 - 5 - 6 794 59
AK094738.1 - 5 - 6 789 59
AK289509.1 - 4 - 5 843 59
AK294748.1 - 5 - 6 662 59
BC013116.1 - 5 - 6 703 59
BC014486.1 - 2 - 3 234 59

View Full Product Details