Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320707.1 | NP_001307636.1 | 12 - 13 | 2229 | 109 |
NM_001320708.1 | NP_001307637.1 | 12 - 13 | 2072 | 109 | |
NM_001320709.1 | NP_001307638.1 | 13 - 14 | 2325 | 109 | |
NM_006256.3 | NP_006247.1 | 13 - 14 | 2373 | 109 | |
XM_011541772.2 | XP_011540074.1 | 8 - 9 | 2711 | 109 | |
XM_017001782.1 | XP_016857271.1 | 12 - 13 | 2138 | 109 | |
XM_017001783.1 | XP_016857272.1 | 12 - 13 | 2132 | 109 | |
GenBank mRNA | AK298595.1 | - | 12 - 13 | 1911 | 109 |
AK300304.1 | - | 13 - 14 | 2224 | 109 | |
AK301066.1 | - | 8 - 9 | 1074 | 109 | |
BC125199.1 | - | 12 - 13 | 1801 | 109 | |
S75548.1 | - | 13 - 14 | 1945 | 109 | |
U33052.1 | - | 13 - 14 | 1945 | 109 |