Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190456.1 | NP_001177385.1 | 6 - 7 | 830 | 78 |
NM_001190457.1 | NP_001177386.1 | 6 - 7 | 1109 | 78 | |
NM_001324014.1 | NP_001310943.1 | 7 - 8 | 1041 | 78 | |
NM_001324015.1 | NP_001310944.1 | 6 - 7 | 842 | 78 | |
NM_006091.4 | NP_006082.3 | 6 - 7 | 1064 | 78 | |
GenBank mRNA | AB010098.1 | - | 6 - 7 | 797 | 78 |
AB023142.1 | - | 5 - 6 | 768 | 78 | |
AK124711.1 | - | 1 - 2 | 1284 | 78 | |
AK289678.1 | - | 6 - 7 | 1109 | 78 | |
BC026335.1 | - | 6 - 7 | 78 |