Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042402.1 | NP_001035861.1 | 1 - 2 | 265 | 82 |
NM_014435.3 | NP_055250.2 | 1 - 2 | 265 | 82 | |
XM_005262920.3 | XP_005262977.1 | 1 - 2 | 263 | 82 | |
XM_005262921.3 | XP_005262978.1 | 1 - 2 | 263 | 82 | |
XM_005262923.2 | XP_005262980.1 | 1 - 2 | 263 | 82 | |
XM_005262924.2 | XP_005262981.1 | 1 - 2 | 263 | 82 | |
XM_005262925.3 | XP_005262982.1 | 1 - 2 | 263 | 82 | |
XM_006714180.2 | XP_006714243.1 | 1 - 2 | 263 | 82 | |
XM_011531852.2 | XP_011530154.1 | 1 - 2 | 263 | 82 | |
XM_011531854.2 | XP_011530156.1 | 1 - 2 | 263 | 82 | |
XM_017008027.1 | XP_016863516.1 | 1 - 2 | 263 | 82 | |
XM_017008028.1 | XP_016863517.1 | 1 - 2 | 263 | 82 | |
XM_017008029.1 | XP_016863518.1 | 1 - 2 | 263 | 82 | |
XM_017008030.1 | XP_016863519.1 | 1 - 2 | 263 | 82 | |
GenBank mRNA | AB161353.1 | - | 1 - 2 | 201 | 82 |
AK024677.1 | - | - | 765 | 82 | |
AK309271.1 | - | 1 - 2 | 237 | 82 | |
BC006388.2 | - | 1 - 2 | 245 | 82 | |
BM763257.1 | - | 1 - 2 | 265 | 82 |