Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290049.1 | NP_001276978.1 | 13 - 14 | 1556 | 82 |
NM_003715.3 | NP_003706.2 | 12 - 13 | 1544 | 82 | |
XM_006714396.3 | XP_006714459.1 | 12 - 13 | 1539 | 82 | |
XM_006714397.3 | XP_006714460.1 | 13 - 14 | 1556 | 82 | |
GenBank mRNA | AK002093.1 | - | 7 - 8 | 764 | 82 |
AK314289.1 | - | 12 - 13 | 1434 | 82 | |
AL832010.2 | - | 13 - 14 | 1479 | 82 | |
AL833753.1 | - | 2 - 3 | 255 | 82 | |
BC006398.2 | - | 12 - 13 | 1469 | 82 | |
BC032654.1 | - | 12 - 13 | 1399 | 82 | |
D86326.1 | - | 12 - 13 | 1421 | 82 |