Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290049.1 | NP_001276978.1 | 6 - 7 | 701 | 66 |
NM_003715.3 | NP_003706.2 | 5 - 6 | 689 | 66 | |
XM_006714396.3 | XP_006714459.1 | 5 - 6 | 684 | 66 | |
XM_006714397.3 | XP_006714460.1 | 6 - 7 | 701 | 66 | |
GenBank mRNA | AK314289.1 | - | 5 - 6 | 579 | 66 |
AL832010.2 | - | 6 - 7 | 624 | 66 | |
BC006398.2 | - | 5 - 6 | 614 | 66 | |
BC032654.1 | - | 5 - 6 | 544 | 66 | |
D86326.1 | - | 5 - 6 | 566 | 66 |