Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030957.3 | NP_112219.3 | 11 - 12 | 1646 | 91 |
XM_006722917.3 | XP_006722980.1 | 10 - 11 | 1492 | 91 | |
XM_017027338.1 | XP_016882827.1 | 10 - 11 | 1502 | 91 | |
XM_017027339.1 | XP_016882828.1 | 1 - 2 | 137 | 91 | |
GenBank mRNA | AB209515.1 | - | 10 - 11 | 1482 | 91 |
BM928283.1 | - | - | 403 | 91 | |
CD632974.1 | - | 5 - 6 | 464 | 90 |