Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004260.3 | NP_004251.3 | 18 - 19 | 3281 | 80 |
XM_011517384.2 | XP_011515686.1 | 18 - 19 | 3083 | 80 | |
XM_017013991.1 | XP_016869480.1 | 16 - 17 | 3451 | 80 | |
XM_017013992.1 | XP_016869481.1 | 17 - 18 | 3375 | 80 | |
XM_017013993.1 | XP_016869482.1 | 17 - 18 | 3360 | 80 | |
XM_017013994.1 | XP_016869483.1 | 17 - 18 | 3356 | 80 | |
XM_017013995.1 | XP_016869484.1 | 18 - 19 | 3283 | 80 | |
XM_017013996.1 | XP_016869485.1 | 16 - 17 | 3449 | 80 | |
XM_017013997.1 | XP_016869486.1 | 16 - 17 | 3253 | 80 | |
XM_017013998.1 | XP_016869487.1 | 17 - 18 | 3373 | 80 | |
XM_017013999.1 | XP_016869488.1 | 17 - 18 | 3162 | 80 | |
XM_017014000.1 | XP_016869489.1 | 12 - 13 | 2362 | 80 | |
XM_017014001.1 | XP_016869490.1 | 15 - 16 | 3452 | 80 | |
GenBank mRNA | AB006532.1 | - | 18 - 19 | 3239 | 80 |
BC011602.2 | - | 11 - 12 | 1840 | 80 | |
BC013277.2 | - | 9 - 10 | 1578 | 80 | |
KJ147518.1 | - | 14 - 15 | 2588 | 80 |