Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004260.3 | NP_004251.3 | 6 - 7 | 1298 | 89 |
XM_011517384.2 | XP_011515686.1 | 6 - 7 | 1298 | 89 | |
XM_017013991.1 | XP_016869480.1 | 6 - 7 | 1303 | 89 | |
XM_017013992.1 | XP_016869481.1 | 6 - 7 | 1302 | 89 | |
XM_017013993.1 | XP_016869482.1 | 6 - 7 | 1302 | 89 | |
XM_017013994.1 | XP_016869483.1 | 7 - 8 | 1208 | 89 | |
XM_017013995.1 | XP_016869484.1 | 6 - 7 | 1300 | 89 | |
XM_017013996.1 | XP_016869485.1 | 6 - 7 | 1301 | 89 | |
XM_017013997.1 | XP_016869486.1 | 6 - 7 | 1303 | 89 | |
XM_017013998.1 | XP_016869487.1 | 6 - 7 | 1300 | 89 | |
XM_017013999.1 | XP_016869488.1 | 6 - 7 | 1302 | 89 | |
GenBank mRNA | AB006532.1 | - | 6 - 7 | 1256 | 89 |
KJ147518.1 | - | 2 - 3 | 803 | 89 |