Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015484.4 | NP_056299.1 | 4 - 5 | 431 | 114 |
NM_207170.3 | NP_997053.1 | 3 - 4 | 305 | 114 | |
GenBank mRNA | AF273089.1 | - | 4 - 5 | 391 | 114 |
AK303582.1 | - | 4 - 5 | 411 | 114 | |
AK314859.1 | - | 4 - 5 | 411 | 114 | |
AL080166.1 | - | 4 - 5 | 392 | 114 | |
BC010862.1 | - | 4 - 5 | 402 | 114 | |
BG390792.1 | - | 3 - 4 | 269 | 114 | |
BI668848.1 | - | 4 - 5 | 429 | 114 | |
CV806249.1 | - | 3 - 4 | 262 | 114 |