Assay Design



Gene Transcripts

Gene Symbol
PPFIA2
Entrez Gene ID
8499
Gene Name
PTPRF interacting protein alpha 2
Gene Aliases
-
Chromosome Location
Chr.12: 81257975 - 81759748 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220473.2 NP_001207402.1 15 - 16 2075 87
NM_001220474.2 NP_001207403.1 14 - 15 2021 87
NM_001220475.2 NP_001207404.1 15 - 16 1887 87
NM_001220476.2 NP_001207405.1 15 - 16 1887 87
NM_001220477.2 NP_001207406.1 13 - 14 1894 87
NM_001220478.2 NP_001207407.1 12 - 13 1819 87
NM_001220479.2 NP_001207408.1 6 - 7 636 87
NM_001220480.2 NP_001207409.1 7 - 8 700 87
NM_001282536.1 NP_001269465.1 13 - 14 1582 87
NM_003625.4 NP_003616.2 16 - 17 1938 87
XM_006719670.3 XP_006719733.1 13 - 14 9130 87
XM_011538906.2 XP_011537208.1 14 - 15 2015 87
XM_011538907.2 XP_011537209.1 14 - 15 2015 87
XM_017020079.1 XP_016875568.1 16 - 17 1922 87
XM_017020080.1 XP_016875569.1 15 - 16 2069 87
XM_017020081.1 XP_016875570.1 15 - 16 2069 87
XM_017020082.1 XP_016875571.1 16 - 17 1922 87
XM_017020083.1 XP_016875572.1 15 - 16 2069 87
XM_017020084.1 XP_016875573.1 15 - 16 2069 87
XM_017020085.1 XP_016875574.1 15 - 16 2069 87
XM_017020086.1 XP_016875575.1 16 - 17 1922 87
XM_017020087.1 XP_016875576.1 15 - 16 2056 87
XM_017020088.1 XP_016875577.1 15 - 16 2302 87
XM_017020089.1 XP_016875578.1 14 - 15 2015 87
XM_017020090.1 XP_016875579.1 16 - 17 1922 87
XM_017020091.1 XP_016875580.1 15 - 16 2069 87
XM_017020092.1 XP_016875581.1 15 - 16 2069 87
XM_017020093.1 XP_016875582.1 15 - 16 2069 87
XM_017020094.1 XP_016875583.1 15 - 16 1847 87
XM_017020095.1 XP_016875584.1 15 - 16 2069 87
XM_017020096.1 XP_016875585.1 14 - 15 1994 87
XM_017020097.1 XP_016875586.1 16 - 17 1922 87
XM_017020098.1 XP_016875587.1 16 - 17 1922 87
XM_017020099.1 XP_016875588.1 14 - 15 2015 87
XM_017020100.1 XP_016875589.1 15 - 16 2069 87
XM_017020101.1 XP_016875590.1 16 - 17 1922 87
XM_017020102.1 XP_016875591.1 14 - 15 1981 87
XM_017020103.1 XP_016875592.1 15 - 16 2069 87
XM_017020104.1 XP_016875593.1 15 - 16 2069 87
XM_017020105.1 XP_016875594.1 14 - 15 1994 87
XM_017020106.1 XP_016875595.1 15 - 16 2069 87
XM_017020107.1 XP_016875596.1 16 - 17 1922 87
XM_017020108.1 XP_016875597.1 16 - 17 1922 87
XM_017020109.1 XP_016875598.1 15 - 16 2069 87
XM_017020110.1 XP_016875599.1 14 - 15 2227 87
XM_017020111.1 XP_016875600.1 15 - 16 2069 87
XM_017020112.1 XP_016875601.1 13 - 14 1894 87
XM_017020113.1 XP_016875602.1 13 - 14 1894 87
XM_017020114.1 XP_016875603.1 13 - 14 1894 87
XM_017020115.1 XP_016875604.1 13 - 14 1894 87
XM_017020116.1 XP_016875605.1 13 - 14 1894 87
XM_017020117.1 XP_016875606.1 12 - 13 1629 87
XM_017020118.1 XP_016875607.1 14 - 15 1869 87
XM_017020119.1 XP_016875608.1 13 - 14 1894 87
XM_017020120.1 XP_016875609.1 13 - 14 1582 87
GenBank mRNA AB210009.1 - 15 - 16 2073 87
AF034799.1 - 16 - 17 1946 87
AK123372.1 - 7 - 8 635 87
AK126971.1 - 14 - 15 1981 87
AK294505.1 - 12 - 13 1629 87
AK296380.1 - 6 - 7 607 87
AK299853.1 - 13 - 14 1843 87
BC104912.1 - 15 - 16 1871 87
BC143485.1 - 15 - 16 1871 87
BU568524.1 - 2 - 3 240 87

View Full Product Details