Assay Design



Gene Transcripts

Gene Symbol
PPFIA2
Entrez Gene ID
8499
Gene Name
PTPRF interacting protein alpha 2
Gene Aliases
-
Chromosome Location
Chr.12: 81257975 - 81759748 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220473.2 NP_001207402.1 21 - 22 2942 71
NM_001220475.2 NP_001207404.1 21 - 22 2754 71
NM_001220476.2 NP_001207405.1 21 - 22 2754 71
NM_001220477.2 NP_001207406.1 19 - 20 2761 71
NM_001220478.2 NP_001207407.1 18 - 19 2686 71
NM_001220479.2 NP_001207408.1 12 - 13 1503 71
NM_001220480.2 NP_001207409.1 13 - 14 1436 71
NM_001282536.1 NP_001269465.1 19 - 20 2449 71
NM_003625.4 NP_003616.2 22 - 23 2805 71
XM_011538906.2 XP_011537208.1 20 - 21 2882 71
XM_011538907.2 XP_011537209.1 20 - 21 2882 71
XM_017020080.1 XP_016875569.1 21 - 22 2936 71
XM_017020082.1 XP_016875571.1 22 - 23 2789 71
XM_017020085.1 XP_016875574.1 21 - 22 2936 71
XM_017020086.1 XP_016875575.1 22 - 23 2789 71
XM_017020090.1 XP_016875579.1 22 - 23 2789 71
XM_017020093.1 XP_016875582.1 21 - 22 2936 71
XM_017020096.1 XP_016875585.1 20 - 21 2861 71
XM_017020097.1 XP_016875586.1 22 - 23 2789 71
XM_017020099.1 XP_016875588.1 20 - 21 2882 71
XM_017020101.1 XP_016875590.1 22 - 23 2789 71
XM_017020102.1 XP_016875591.1 20 - 21 2848 71
XM_017020103.1 XP_016875592.1 21 - 22 2936 71
XM_017020105.1 XP_016875594.1 20 - 21 2861 71
XM_017020106.1 XP_016875595.1 21 - 22 2936 71
XM_017020107.1 XP_016875596.1 22 - 23 2789 71
XM_017020109.1 XP_016875598.1 21 - 22 2936 71
XM_017020110.1 XP_016875599.1 20 - 21 3094 71
XM_017020111.1 XP_016875600.1 21 - 22 2936 71
XM_017020112.1 XP_016875601.1 19 - 20 2761 71
XM_017020113.1 XP_016875602.1 19 - 20 2761 71
XM_017020115.1 XP_016875604.1 19 - 20 2761 71
XM_017020116.1 XP_016875605.1 19 - 20 2761 71
XM_017020119.1 XP_016875608.1 19 - 20 2761 71
GenBank mRNA AB210009.1 - 21 - 22 2940 71
AF034799.1 - 22 - 23 2813 71
AK123372.1 - 13 - 14 1371 71
AK294505.1 - 18 - 19 2496 71
AK296380.1 - 12 - 13 1474 71
AK299853.1 - 19 - 20 2710 71
AK302658.1 - 19 - 20 2420 71
BC104912.1 - 21 - 22 2738 71
BC143485.1 - 21 - 22 2738 71

View Full Product Details