Assay Design



Gene Transcripts

Gene Symbol
PPFIA2
Entrez Gene ID
8499
Gene Name
PTPRF interacting protein alpha 2
Gene Aliases
-
Chromosome Location
Chr.12: 81257975 - 81759748 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220473.2 NP_001207402.1 1 - 2 298 64
NM_001220474.2 NP_001207403.1 1 - 2 298 64
NM_001220475.2 NP_001207404.1 1 - 2 110 64
NM_001220476.2 NP_001207405.1 1 - 2 110 64
NM_003625.4 NP_003616.2 2 - 3 161 64
XM_011538906.2 XP_011537208.1 1 - 2 292 64
XM_011538907.2 XP_011537209.1 1 - 2 292 64
XM_017020079.1 XP_016875568.1 2 - 3 145 64
XM_017020080.1 XP_016875569.1 1 - 2 292 64
XM_017020081.1 XP_016875570.1 1 - 2 292 64
XM_017020082.1 XP_016875571.1 2 - 3 145 64
XM_017020083.1 XP_016875572.1 1 - 2 292 64
XM_017020084.1 XP_016875573.1 1 - 2 292 64
XM_017020085.1 XP_016875574.1 1 - 2 292 64
XM_017020086.1 XP_016875575.1 2 - 3 145 64
XM_017020087.1 XP_016875576.1 1 - 2 279 64
XM_017020088.1 XP_016875577.1 2 - 3 579 64
XM_017020089.1 XP_016875578.1 1 - 2 292 64
XM_017020090.1 XP_016875579.1 2 - 3 145 64
XM_017020091.1 XP_016875580.1 1 - 2 292 64
XM_017020092.1 XP_016875581.1 1 - 2 292 64
XM_017020093.1 XP_016875582.1 1 - 2 292 64
XM_017020094.1 XP_016875583.1 2 - 3 145 64
XM_017020095.1 XP_016875584.1 1 - 2 292 64
XM_017020096.1 XP_016875585.1 1 - 2 292 64
XM_017020097.1 XP_016875586.1 2 - 3 145 64
XM_017020098.1 XP_016875587.1 2 - 3 145 64
XM_017020099.1 XP_016875588.1 1 - 2 292 64
XM_017020100.1 XP_016875589.1 1 - 2 292 64
XM_017020101.1 XP_016875590.1 2 - 3 145 64
XM_017020102.1 XP_016875591.1 1 - 2 279 64
XM_017020103.1 XP_016875592.1 1 - 2 292 64
XM_017020104.1 XP_016875593.1 1 - 2 292 64
XM_017020105.1 XP_016875594.1 1 - 2 292 64
XM_017020106.1 XP_016875595.1 1 - 2 292 64
XM_017020107.1 XP_016875596.1 2 - 3 145 64
XM_017020108.1 XP_016875597.1 2 - 3 145 64
XM_017020109.1 XP_016875598.1 1 - 2 292 64
XM_017020110.1 XP_016875599.1 2 - 3 579 64
XM_017020111.1 XP_016875600.1 1 - 2 292 64
GenBank mRNA AB210009.1 - 1 - 2 296 64
AF034799.1 - 2 - 3 169 64
AK126971.1 - 1 - 2 258 64
AK307524.1 - 2 - 3 145 64
BC104912.1 - 1 - 2 94 64
BC143485.1 - 1 - 2 94 64
DA499362.1 - 1 - 2 299 64

View Full Product Details