Assay Design



Gene Transcripts

Gene Symbol
PPFIA2
Entrez Gene ID
8499
Gene Name
PTPRF interacting protein alpha 2
Gene Aliases
-
Chromosome Location
Chr.12: 81257975 - 81759748 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220473.2 NP_001207402.1 29 - 30 3849 65
NM_001220474.2 NP_001207403.1 29 - 30 3804 65
NM_001220475.2 NP_001207404.1 28 - 29 3598 65
NM_001220476.2 NP_001207405.1 29 - 30 3643 65
NM_001220477.2 NP_001207406.1 26 - 27 3587 65
NM_001220478.2 NP_001207407.1 26 - 27 3575 65
NM_001220479.2 NP_001207408.1 18 - 19 2317 65
NM_001220480.2 NP_001207409.1 19 - 20 2250 65
NM_001282536.1 NP_001269465.1 27 - 28 3356 65
NM_003625.4 NP_003616.2 30 - 31 3712 65
XM_006719670.3 XP_006719733.1 28 - 29 10913 65
XM_011538906.2 XP_011537208.1 26 - 27 3678 65
XM_011538907.2 XP_011537209.1 28 - 29 3771 65
XM_017020079.1 XP_016875568.1 31 - 32 3705 65
XM_017020080.1 XP_016875569.1 29 - 30 3843 65
XM_017020081.1 XP_016875570.1 30 - 31 3834 65
XM_017020082.1 XP_016875571.1 30 - 31 3678 65
XM_017020083.1 XP_016875572.1 29 - 30 3822 65
XM_017020084.1 XP_016875573.1 30 - 31 3852 65
XM_017020085.1 XP_016875574.1 28 - 29 3813 65
XM_017020086.1 XP_016875575.1 30 - 31 3696 65
XM_017020087.1 XP_016875576.1 30 - 31 3821 65
XM_017020088.1 XP_016875577.1 30 - 31 4085 65
XM_017020089.1 XP_016875578.1 29 - 30 3798 65
XM_017020090.1 XP_016875579.1 30 - 31 3678 65
XM_017020091.1 XP_016875580.1 29 - 30 3822 65
XM_017020092.1 XP_016875581.1 29 - 30 3789 65
XM_017020093.1 XP_016875582.1 28 - 29 3813 65
XM_017020094.1 XP_016875583.1 30 - 31 3630 65
XM_017020095.1 XP_016875584.1 29 - 30 3771 65
XM_017020096.1 XP_016875585.1 28 - 29 3768 65
XM_017020097.1 XP_016875586.1 29 - 30 3615 65
XM_017020098.1 XP_016875587.1 29 - 30 3612 65
XM_017020099.1 XP_016875588.1 28 - 29 3789 65
XM_017020100.1 XP_016875589.1 29 - 30 3789 65
XM_017020101.1 XP_016875590.1 28 - 29 3603 65
XM_017020102.1 XP_016875591.1 28 - 29 3737 65
XM_017020103.1 XP_016875592.1 28 - 29 3780 65
XM_017020104.1 XP_016875593.1 28 - 29 3741 65
XM_017020105.1 XP_016875594.1 28 - 29 3768 65
XM_017020106.1 XP_016875595.1 27 - 28 3732 65
XM_017020107.1 XP_016875596.1 29 - 30 3615 65
XM_017020108.1 XP_016875597.1 29 - 30 3612 65
XM_017020109.1 XP_016875598.1 27 - 28 3750 65
XM_017020110.1 XP_016875599.1 28 - 29 4001 65
XM_017020111.1 XP_016875600.1 27 - 28 3732 65
XM_017020112.1 XP_016875601.1 27 - 28 3668 65
XM_017020113.1 XP_016875602.1 27 - 28 3650 65
XM_017020114.1 XP_016875603.1 28 - 29 3677 65
XM_017020115.1 XP_016875604.1 27 - 28 3668 65
XM_017020116.1 XP_016875605.1 27 - 28 3650 65
XM_017020117.1 XP_016875606.1 27 - 28 3412 65
XM_017020118.1 XP_016875607.1 29 - 30 3652 65
XM_017020119.1 XP_016875608.1 26 - 27 3605 65
XM_017020120.1 XP_016875609.1 28 - 29 3365 65
GenBank mRNA AB210009.1 - 29 - 30 3847 65
AF034799.1 - 30 - 31 3720 65
AK126971.1 - 29 - 30 3764 65
AK294505.1 - 26 - 27 3385 65
AK296380.1 - 18 - 19 2288 65
AK299853.1 - 26 - 27 3536 65
AK302658.1 - 27 - 28 3327 65
BC104912.1 - 28 - 29 3582 65
BC143485.1 - 29 - 30 3627 65
BM689808.1 - 2 - 3 226 65

View Full Product Details