Assay Design



Gene Transcripts

Gene Symbol
PPFIA2
Entrez Gene ID
8499
Gene Name
PTPRF interacting protein alpha 2
Gene Aliases
-
Chromosome Location
Chr.12: 81257975 - 81759748 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001220475.2 NP_001207404.1 29 - 30 3759 69
NM_001220476.2 NP_001207405.1 30 - 31 3804 69
NM_001220477.2 NP_001207406.1 27 - 28 3748 69
NM_001220478.2 NP_001207407.1 27 - 28 3736 69
NM_001220480.2 NP_001207409.1 20 - 21 2411 69
NM_001282536.1 NP_001269465.1 28 - 29 3517 69
NM_003625.4 NP_003616.2 31 - 32 3873 69
XM_006719670.3 XP_006719733.1 29 - 30 11074 69
XM_011538906.2 XP_011537208.1 27 - 28 3839 69
XM_011538907.2 XP_011537209.1 29 - 30 3932 69
XM_017020079.1 XP_016875568.1 32 - 33 3866 69
XM_017020080.1 XP_016875569.1 30 - 31 4004 69
XM_017020081.1 XP_016875570.1 31 - 32 3995 69
XM_017020082.1 XP_016875571.1 31 - 32 3839 69
XM_017020083.1 XP_016875572.1 30 - 31 3983 69
XM_017020085.1 XP_016875574.1 29 - 30 3974 69
XM_017020088.1 XP_016875577.1 31 - 32 4246 69
XM_017020089.1 XP_016875578.1 30 - 31 3959 69
XM_017020092.1 XP_016875581.1 30 - 31 3950 69
XM_017020094.1 XP_016875583.1 31 - 32 3791 69
XM_017020095.1 XP_016875584.1 30 - 31 3932 69
XM_017020096.1 XP_016875585.1 29 - 30 3929 69
XM_017020097.1 XP_016875586.1 30 - 31 3776 69
XM_017020098.1 XP_016875587.1 30 - 31 3773 69
XM_017020101.1 XP_016875590.1 29 - 30 3764 69
XM_017020102.1 XP_016875591.1 29 - 30 3898 69
XM_017020104.1 XP_016875593.1 29 - 30 3902 69
XM_017020106.1 XP_016875595.1 28 - 29 3893 69
XM_017020110.1 XP_016875599.1 29 - 30 4162 69
XM_017020112.1 XP_016875601.1 28 - 29 3829 69
XM_017020113.1 XP_016875602.1 28 - 29 3811 69
XM_017020117.1 XP_016875606.1 28 - 29 3573 69
XM_017020118.1 XP_016875607.1 30 - 31 3813 69
XM_017020119.1 XP_016875608.1 27 - 28 3766 69
XM_017020120.1 XP_016875609.1 29 - 30 3526 69
GenBank mRNA AF034799.1 - 31 - 32 3881 69
AK123372.1 - 20 - 21 2346 69
AK294505.1 - 27 - 28 3546 69
AK299853.1 - 27 - 28 3697 69
AK302658.1 - 28 - 29 3488 69
BC104912.1 - 29 - 30 3743 69
BC143485.1 - 30 - 31 3788 69
BM689808.1 - 3 - 4 387 69

View Full Product Details