Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032880.4 | NP_116269.3 | 6 - 7 | 1397 | 68 |
XM_017002604.1 | XP_016858093.1 | 6 - 7 | 1043 | 68 | |
XM_017002605.1 | XP_016858094.1 | 5 - 6 | 860 | 68 | |
GenBank mRNA | AK075316.1 | - | 6 - 7 | 1397 | 68 |
AK123172.1 | - | 2 - 3 | 1338 | 68 | |
AK294121.1 | - | 6 - 7 | 966 | 68 | |
BC007618.1 | - | 6 - 7 | 1317 | 68 |