Assay Design
Gene Transcripts
- Gene Symbol
- CELF1
- Entrez Gene ID
-
10658
- Gene Name
- CUGBP, Elav-like family member 1
- Gene Aliases
-
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
- Chromosome Location
-
Chr.11: 47465937 - 47565569 on Build GRCh38
View Full Product Details