Assay Design



Gene Transcripts

Gene Symbol
DGCR14
Entrez Gene ID
8220
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el
Chromosome Location
Chr.22: 19130279 - 19144726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022719.2 NP_073210.1 10 - 10 2604 67
NR_134304.1 10 - 10 2681 67
GenBank mRNA BC069275.1 - 1 - 1 241 67
L77565.1 - 1 - 1 639 67

View Full Product Details