Assay Design



Gene Transcripts

Gene Symbol
KLK12
Entrez Gene ID
43849
Gene Name
kallikrein related peptidase 12
Gene Aliases
KLK-L5, KLKL5
Chromosome Location
Chr.19: 51028954 - 51035651 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019598.2 NP_062544.1 2 - 3 151 129
NM_145894.1 NP_665901.1 2 - 3 151 129
NM_145895.1 NP_665902.1 2 - 3 151 129

View Full Product Details