Assay Design



Gene Transcripts

Gene Symbol
KRT1
Entrez Gene ID
3848
Gene Name
keratin 1
Gene Aliases
CK1, EHK, EHK1, EPPK, K1, KRT1A, NEPPK
Chromosome Location
Chr.12: 52674736 - 52680407 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006121.3 NP_006112.3 4 - 5 1021 127
GenBank mRNA AK313986.1 - 4 - 5 1021 127
BC063697.1 - 4 - 5 1003 127

View Full Product Details