Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077261.3 | NP_001070729.2 | 28 - 29 | 3986 | 62 |
NM_001206654.1 | NP_001193583.1 | 28 - 29 | 3986 | 62 | |
NM_006312.5 | NP_006303.4 | 29 - 30 | 4016 | 62 | |
GenBank mRNA | AB209089.1 | - | 29 - 30 | 4051 | 62 |
AF113003.1 | - | 28 - 29 | 3696 | 62 | |
AF125672.1 | - | 27 - 28 | 3821 | 62 | |
AY965853.1 | - | 27 - 28 | 3821 | 62 |