Assay Design



Gene Transcripts

Gene Symbol
NCOR2
Entrez Gene ID
9612
Gene Name
nuclear receptor corepressor 2
Gene Aliases
CTG26, N-CoR2, SMAP270, SMRT, SMRTE, SMRTE-tau, TNRC14, TRAC, TRAC-1, TRAC1
Chromosome Location
Chr.12: 124324411 - 124567464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077261.3 NP_001070729.2 41 - 42 6698 127
NM_001206654.1 NP_001193583.1 41 - 42 6698 127
NM_006312.5 NP_006303.4 42 - 43 6728 127
GenBank mRNA AF113003.1 - 41 - 42 6417 127
AF125672.1 - 40 - 41 6542 127
AY965853.1 - 40 - 41 6542 127
BC004326.1 - 3 - 4 683 127
S83390.1 - 8 - 9 1765 127

View Full Product Details