Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306129.1 | NP_001293058.1 | 10 - 11 | 1809 | 114 |
NM_001306130.1 | NP_001293059.1 | 10 - 11 | 1809 | 114 | |
NM_001306131.1 | NP_001293060.1 | 10 - 11 | 1809 | 114 | |
NM_001306132.1 | NP_001293061.1 | 10 - 11 | 1809 | 114 | |
NM_002026.3 | NP_002017.1 | 10 - 11 | 1807 | 114 | |
NM_054034.2 | NP_473375.2 | 10 - 11 | 1807 | 114 | |
NM_212474.2 | NP_997639.1 | 10 - 11 | 1807 | 114 | |
NM_212476.2 | NP_997641.1 | 10 - 11 | 1807 | 114 | |
NM_212478.2 | NP_997643.1 | 10 - 11 | 1807 | 114 | |
NM_212482.2 | NP_997647.1 | 10 - 11 | 1807 | 114 | |
XM_005246397.1 | XP_005246454.1 | 10 - 11 | 1906 | 114 | |
XM_005246398.1 | XP_005246455.1 | 10 - 11 | 1906 | 114 | |
XM_005246399.1 | XP_005246456.1 | 10 - 11 | 1906 | 114 | |
XM_005246401.1 | XP_005246458.1 | 10 - 11 | 1906 | 114 | |
XM_005246402.1 | XP_005246459.1 | 10 - 11 | 1906 | 114 | |
XM_005246403.1 | XP_005246460.1 | 10 - 11 | 1906 | 114 | |
XM_005246404.1 | XP_005246461.1 | 10 - 11 | 1906 | 114 | |
XM_005246405.1 | XP_005246462.1 | 10 - 11 | 1906 | 114 | |
XM_005246406.1 | XP_005246463.1 | 10 - 11 | 1906 | 114 | |
XM_005246407.1 | XP_005246464.1 | 10 - 11 | 1906 | 114 | |
XM_005246408.1 | XP_005246465.1 | 10 - 11 | 1906 | 114 | |
XM_005246409.1 | XP_005246466.1 | 10 - 11 | 1906 | 114 | |
XM_005246410.1 | XP_005246467.1 | 10 - 11 | 1906 | 114 | |
XM_005246411.1 | XP_005246468.1 | 10 - 11 | 1906 | 114 | |
XM_005246412.1 | XP_005246469.1 | 10 - 11 | 1906 | 114 | |
XM_005246414.1 | XP_005246471.1 | 10 - 11 | 1906 | 114 | |
XM_005246416.1 | XP_005246473.1 | 10 - 11 | 1906 | 114 | |
XM_017003692.1 | XP_016859181.1 | 10 - 11 | 1906 | 114 | |
XM_017003693.1 | XP_016859182.1 | 10 - 11 | 1906 | 114 | |
XM_017003694.1 | XP_016859183.1 | 10 - 11 | 1906 | 114 | |
XM_017003695.1 | XP_016859184.1 | 10 - 11 | 1906 | 114 | |
GenBank mRNA | AB191261.1 | - | 10 - 11 | 1807 | 114 |
AB209840.1 | - | 10 - 11 | 1799 | 114 | |
AJ276395.1 | - | 9 - 10 | 1552 | 114 | |
AJ535086.1 | - | 10 - 11 | 1597 | 114 | |
AJ849445.1 | - | 10 - 11 | 1597 | 114 | |
AL832202.1 | - | 10 - 11 | 1808 | 114 | |
AL832771.1 | - | 10 - 11 | 1808 | 114 | |
BC117176.1 | - | 10 - 11 | 1787 | 114 | |
BC143754.1 | - | 10 - 11 | 1787 | 114 | |
BC143763.1 | - | 10 - 11 | 1787 | 114 | |
BX538017.1 | - | 10 - 11 | 1808 | 114 | |
BX538018.1 | - | 10 - 11 | 1808 | 114 | |
BX538045.1 | - | 10 - 11 | 1847 | 114 | |
BX640608.1 | - | 10 - 11 | 1807 | 114 | |
BX640731.1 | - | 10 - 11 | 1808 | 114 | |
BX640802.1 | - | 10 - 11 | 1810 | 114 | |
BX640803.1 | - | 10 - 11 | 1807 | 114 | |
BX640875.1 | - | 10 - 11 | 1808 | 114 | |
BX640999.1 | - | 5 - 6 | 783 | 114 | |
BX649182.1 | - | 10 - 11 | 1810 | 114 | |
CR749281.1 | - | 10 - 11 | 1846 | 114 | |
CR749316.1 | - | 10 - 11 | 1806 | 114 | |
CR749317.1 | - | 10 - 11 | 1806 | 114 |