Assay Design



Gene Transcripts

Gene Symbol
FN1
Entrez Gene ID
2335
Gene Name
fibronectin 1
Gene Aliases
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
Chromosome Location
Chr.2: 215360440 - 215436167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306129.1 NP_001293058.1 3 - 4 681 73
NM_001306130.1 NP_001293059.1 3 - 4 681 73
NM_001306131.1 NP_001293060.1 3 - 4 681 73
NM_001306132.1 NP_001293061.1 3 - 4 681 73
NM_002026.3 NP_002017.1 3 - 4 679 73
NM_054034.2 NP_473375.2 3 - 4 679 73
NM_212474.2 NP_997639.1 3 - 4 679 73
NM_212476.2 NP_997641.1 3 - 4 679 73
NM_212478.2 NP_997643.1 3 - 4 679 73
NM_212482.2 NP_997647.1 3 - 4 679 73
XM_005246397.1 XP_005246454.1 3 - 4 778 73
XM_005246398.1 XP_005246455.1 3 - 4 778 73
XM_005246399.1 XP_005246456.1 3 - 4 778 73
XM_005246401.1 XP_005246458.1 3 - 4 778 73
XM_005246402.1 XP_005246459.1 3 - 4 778 73
XM_005246403.1 XP_005246460.1 3 - 4 778 73
XM_005246404.1 XP_005246461.1 3 - 4 778 73
XM_005246405.1 XP_005246462.1 3 - 4 778 73
XM_005246406.1 XP_005246463.1 3 - 4 778 73
XM_005246407.1 XP_005246464.1 3 - 4 778 73
XM_005246408.1 XP_005246465.1 3 - 4 778 73
XM_005246409.1 XP_005246466.1 3 - 4 778 73
XM_005246410.1 XP_005246467.1 3 - 4 778 73
XM_005246411.1 XP_005246468.1 3 - 4 778 73
XM_005246412.1 XP_005246469.1 3 - 4 778 73
XM_005246414.1 XP_005246471.1 3 - 4 778 73
XM_005246416.1 XP_005246473.1 3 - 4 778 73
XM_017003692.1 XP_016859181.1 3 - 4 778 73
XM_017003693.1 XP_016859182.1 3 - 4 778 73
XM_017003694.1 XP_016859183.1 3 - 4 778 73
XM_017003695.1 XP_016859184.1 3 - 4 778 73
GenBank mRNA AB191261.1 - 3 - 4 679 73
AB209840.1 - 3 - 4 671 73
AJ276395.1 - 3 - 4 469 73
AJ535086.1 - 3 - 4 469 73
AJ849445.1 - 3 - 4 469 73
AL832202.1 - 3 - 4 680 73
AL832771.1 - 3 - 4 680 73
BC117176.1 - 3 - 4 659 73
BC143754.1 - 3 - 4 659 73
BC143763.1 - 3 - 4 659 73
BX537590.1 - 3 - 4 680 73
BX538017.1 - 3 - 4 680 73
BX538018.1 - 3 - 4 680 73
BX538045.1 - 3 - 4 719 73
BX640608.1 - 3 - 4 679 73
BX640731.1 - 3 - 4 680 73
BX640802.1 - 3 - 4 682 73
BX640803.1 - 3 - 4 679 73
BX640875.1 - 3 - 4 680 73
BX649182.1 - 3 - 4 682 73
CR749281.1 - 3 - 4 718 73
CR749316.1 - 3 - 4 678 73
CR749317.1 - 3 - 4 678 73
U42404.1 - 1 - 2 108 73
X02761.1 - 3 - 4 333 73

View Full Product Details