Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015711.3 | NP_056526.3 | 8 - 9 | 3091 | 112 |
XM_005258833.4 | XP_005258890.1 | 8 - 9 | 3134 | 112 | |
XM_011526882.2 | XP_011525184.1 | 4 - 5 | 4101 | 112 | |
XM_011526883.2 | XP_011525185.1 | 7 - 8 | 2957 | 112 | |
GenBank mRNA | AF182077.1 | - | 3 - 4 | 3107 | 112 |