Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015711.3 | NP_056526.3 | 9 - 10 | 3273 | 101 |
XM_005258833.4 | XP_005258890.1 | 9 - 10 | 3316 | 101 | |
XM_011526882.2 | XP_011525184.1 | 5 - 6 | 4283 | 101 | |
XM_011526883.2 | XP_011525185.1 | 8 - 9 | 3139 | 101 | |
GenBank mRNA | AF182077.1 | - | 4 - 5 | 3289 | 101 |