Assay Design



Gene Transcripts

Gene Symbol
GLTSCR1
Entrez Gene ID
29998
Gene Name
glioma tumor suppressor candidate region gene 1
Gene Aliases
-
Chromosome Location
Chr.19: 47608196 - 47703277 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015711.3 NP_056526.3 12 - 13 3590 108
XM_005258833.4 XP_005258890.1 12 - 13 3633 108
XM_006723180.3 XP_006723243.1 8 - 9 2553 108
XM_011526882.2 XP_011525184.1 8 - 9 4600 108
XM_011526883.2 XP_011525185.1 11 - 12 3456 108
GenBank mRNA AF182077.1 - 7 - 8 3606 108

View Full Product Details