Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015711.3 | NP_056526.3 | 14 - 15 | 3785 | 67 |
XM_005258833.4 | XP_005258890.1 | 14 - 15 | 3828 | 67 | |
XM_006723180.3 | XP_006723243.1 | 10 - 11 | 2748 | 67 | |
XM_011526882.2 | XP_011525184.1 | 10 - 11 | 4795 | 67 | |
XM_011526883.2 | XP_011525185.1 | 13 - 14 | 3651 | 67 | |
GenBank mRNA | AF182077.1 | - | 9 - 10 | 3801 | 67 |