Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005455.4 | NP_005446.2 | 5 - 6 | 671 | 128 |
NM_203350.2 | NP_976225.1 | 5 - 6 | 671 | 128 | |
GenBank mRNA | AB209568.1 | - | 5 - 6 | 384 | 128 |
AF065391.1 | - | 5 - 6 | 411 | 128 | |
AF065392.1 | - | 5 - 6 | 411 | 128 | |
AK023252.1 | - | 2 - 3 | 150 | 128 | |
AK222858.1 | - | 5 - 6 | 396 | 128 | |
AL136945.1 | - | 5 - 6 | 393 | 128 | |
BC033648.1 | - | 5 - 6 | 417 | 128 | |
BC039814.1 | - | 5 - 6 | 417 | 128 |