Assay Design



Gene Transcripts

Gene Symbol
PARPBP
Entrez Gene ID
55010
Gene Name
PARP1 binding protein
Gene Aliases
AROM, C12orf48, PARI
Chromosome Location
Chr.12: 102120171 - 102197520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319988.1 NP_001306917.1 1 - 2 111 122
NM_001319993.1 NP_001306922.1 1 - 2 111 122
NM_001319994.1 NP_001306923.1 1 - 2 111 122
NM_001319995.1 NP_001306924.1 1 - 2 111 122
NM_001319996.1 NP_001306925.1 1 - 2 111 122
NM_017915.4 NP_060385.3 1 - 2 111 122
NR_135123.1 1 - 2 111 122
XM_011538509.1 XP_011536811.1 1 - 2 111 122
XM_017019534.1 XP_016875023.1 1 - 2 111 122
XM_017019536.1 XP_016875025.1 1 - 2 111 122
XM_017019537.1 XP_016875026.1 1 - 2 111 122
XM_017019538.1 XP_016875027.1 1 - 2 111 122
XM_017019539.1 XP_016875028.1 1 - 2 111 122
XM_017019540.1 XP_016875029.1 1 - 2 78 122
XM_017019541.1 XP_016875030.1 1 - 2 111 122
XM_017019542.1 XP_016875031.1 1 - 2 111 122
XM_017019543.1 XP_016875032.1 1 - 2 105 122
XM_017019544.1 XP_016875033.1 1 - 2 105 122
GenBank mRNA AF274940.1 - 1 - 2 60 123
AK300041.1 - 1 - 2 87 122
AK302724.1 - 1 - 2 97 122
AK303571.1 - 1 - 2 37 122
BC018903.2 - 1 - 2 53 122
BC030962.1 - 1 - 2 55 122
BC070270.1 - 1 - 2 67 122
DB260405.1 - 1 - 2 111 122

View Full Product Details