Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287516.1 | NP_001274445.1 | 11 - 12 | 1358 | 67 |
NM_001287517.1 | NP_001274446.1 | 11 - 12 | 1316 | 67 | |
NM_001287518.1 | NP_001274447.1 | 10 - 11 | 1277 | 67 | |
NM_001287519.1 | NP_001274448.1 | 11 - 12 | 1089 | 67 | |
NM_001287520.1 | NP_001274449.1 | 11 - 12 | 1131 | 67 | |
NM_001287522.1 | NP_001274451.1 | 10 - 11 | 1008 | 67 | |
NM_001287524.1 | NP_001274453.1 | 7 - 8 | 913 | 67 | |
NM_004358.4 | NP_004349.1 | 11 - 12 | 1941 | 67 | |
NM_021872.3 | NP_068658.1 | 10 - 11 | 1860 | 67 | |
NM_021873.3 | NP_068659.1 | 11 - 12 | 1983 | 67 | |
GenBank mRNA | AK092713.1 | - | 11 - 12 | 1131 | 67 |
AK295573.1 | - | 11 - 12 | 1200 | 67 | |
AK299028.1 | - | 10 - 11 | 1084 | 67 | |
AK299192.1 | - | 11 - 12 | 1134 | 67 | |
BC006395.2 | - | 11 - 12 | 1264 | 67 | |
BC009953.2 | - | 11 - 12 | 1344 | 67 | |
BC051711.1 | - | 11 - 12 | 1888 | 67 | |
BX640836.1 | - | 7 - 8 | 905 | 67 | |
BX647988.1 | - | 11 - 12 | 1046 | 67 | |
FR695900.1 | - | 11 - 12 | 1131 | 67 | |
FR695901.1 | - | 10 - 11 | 1008 | 67 | |
M81934.1 | - | 11 - 12 | 1220 | 67 | |
S78187.1 | - | 11 - 12 | 1388 | 67 | |
Z68092.1 | - | 10 - 11 | 1168 | 67 |