Assay Design
Gene Transcripts
- Gene Symbol
- HIST1H2BB
- Entrez Gene ID
-
3018
- Gene Name
- histone cluster 1, H2bb
- Gene Aliases
-
H2B.1, H2B/f, H2BFF
- Chromosome Location
-
Chr.6: 26043227 - 26043657 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_021062.2 |
NP_066406.1 |
1 - 1 |
57 |
77 |
GenBank mRNA |
BC096728.1 |
- |
1 - 1 |
57 |
77 |
View Full Product Details