Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101421.3 | NP_001094891.3 | 16 - 17 | 1680 | 117 |
XM_011538223.2 | XP_011536525.1 | 16 - 17 | 2122 | 117 | |
XM_017019207.1 | XP_016874696.1 | 16 - 17 | 2122 | 117 | |
GenBank mRNA | CR747253.1 | - | 2 - 3 | 108 | 117 |
DV080385.1 | - | 4 - 5 | 245 | 117 |