Assay Design



Gene Transcripts

Gene Symbol
MYO1H
Entrez Gene ID
283446
Gene Name
myosin IH
Gene Aliases
-
Chromosome Location
Chr.12: 109388277 - 109448371 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101421.3 NP_001094891.3 8 - 9 993 102
XM_011538223.2 XP_011536525.1 8 - 9 1435 102
XM_017019207.1 XP_016874696.1 8 - 9 1435 102

View Full Product Details