Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006914.3 | NP_008845.2 | 6 - 7 | 1534 | 83 |
GenBank mRNA | AB307695.1 | - | 5 - 6 | 893 | 83 |
AK313431.1 | - | 6 - 7 | 1202 | 83 | |
BC051830.1 | - | 6 - 7 | 1522 | 83 | |
BC093772.1 | - | 6 - 7 | 968 | 83 | |
BC093774.1 | - | 6 - 7 | 968 | 83 | |
BX647070.1 | - | 6 - 7 | 975 | 83 | |
HQ709186.1 | - | 5 - 6 | 893 | 83 | |
Y08639.1 | - | 6 - 7 | 1487 | 83 |