Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005366.1 | NP_001005366.1 | 12 - 13 | 2054 | 69 |
NM_032590.4 | NP_115979.3 | 12 - 13 | 1844 | 69 | |
XM_005253955.3 | XP_005254012.1 | 12 - 13 | 2054 | 69 | |
XM_005253956.3 | XP_005254013.1 | 12 - 13 | 1651 | 69 | |
XM_011538867.2 | XP_011537169.1 | 12 - 13 | 3097 | 69 | |
XM_011538868.2 | XP_011537170.1 | 12 - 13 | 3093 | 69 | |
XM_011538869.2 | XP_011537171.1 | 12 - 13 | 3095 | 69 | |
GenBank mRNA | AB031230.1 | - | 2 - 3 | 303 | 69 |
AJ459424.1 | - | 12 - 13 | 1816 | 69 | |
AK027440.1 | - | 2 - 3 | 233 | 69 | |
AK127328.1 | - | 12 - 13 | 2054 | 69 | |
AL133572.1 | - | 8 - 9 | 1357 | 69 | |
BC115379.1 | - | 11 - 12 | 1704 | 69 | |
BC115380.1 | - | 12 - 13 | 1783 | 69 |