Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005366.1 | NP_001005366.1 | 20 - 21 | 3812 | 60 |
NM_032590.4 | NP_115979.3 | 21 - 22 | 3716 | 60 | |
XM_005253955.3 | XP_005254012.1 | 21 - 22 | 3926 | 60 | |
XM_005253956.3 | XP_005254013.1 | 21 - 22 | 3523 | 60 | |
XM_005253961.4 | XP_005254018.1 | 10 - 11 | 2011 | 60 | |
XM_011538867.2 | XP_011537169.1 | 21 - 22 | 4978 | 60 | |
XM_011538868.2 | XP_011537170.1 | 21 - 22 | 4974 | 60 | |
XM_011538869.2 | XP_011537171.1 | 21 - 22 | 4976 | 60 | |
XM_011538875.2 | XP_011537177.1 | 10 - 11 | 2020 | 60 | |
GenBank mRNA | AB031230.1 | - | 11 - 12 | 2184 | 60 |
AJ459424.1 | - | 21 - 22 | 3688 | 60 | |
AK027440.1 | - | 11 - 12 | 2105 | 60 | |
AK027692.1 | - | 8 - 9 | 1621 | 60 | |
AK074718.1 | - | 6 - 7 | 1200 | 60 | |
AK127328.1 | - | 20 - 21 | 3812 | 60 | |
AK299827.1 | - | 10 - 11 | 2030 | 60 | |
AK316470.1 | - | 10 - 11 | 2023 | 60 | |
BC008735.2 | - | 7 - 8 | 1486 | 60 |