Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005366.1 | NP_001005366.1 | 21 - 22 | 4033 | 69 |
NM_032590.4 | NP_115979.3 | 22 - 23 | 3937 | 69 | |
XM_005253955.3 | XP_005254012.1 | 22 - 23 | 4147 | 69 | |
XM_005253956.3 | XP_005254013.1 | 22 - 23 | 3744 | 69 | |
XM_005253961.4 | XP_005254018.1 | 11 - 12 | 2232 | 69 | |
XM_011538867.2 | XP_011537169.1 | 22 - 23 | 5199 | 69 | |
XM_011538868.2 | XP_011537170.1 | 22 - 23 | 5195 | 69 | |
XM_011538869.2 | XP_011537171.1 | 22 - 23 | 5197 | 69 | |
XM_011538875.2 | XP_011537177.1 | 11 - 12 | 2241 | 69 | |
GenBank mRNA | AJ459424.1 | - | 22 - 23 | 3909 | 69 |
AK027440.1 | - | 12 - 13 | 2326 | 69 | |
AK027692.1 | - | 9 - 10 | 1842 | 69 | |
AK074718.1 | - | 7 - 8 | 1421 | 69 | |
AK127328.1 | - | 21 - 22 | 4033 | 69 | |
AK299827.1 | - | 11 - 12 | 2251 | 69 | |
AK316470.1 | - | 11 - 12 | 2244 | 69 | |
AL133572.1 | - | 14 - 15 | 5732 | 69 | |
BC008735.2 | - | 8 - 9 | 1707 | 69 |