Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000545.6 | NP_000536.5 | 5 - 6 | 1305 | 89 |
NM_001306179.1 | NP_001293108.1 | 5 - 6 | 1305 | 89 | |
XM_005253931.3 | XP_005253988.1 | 5 - 6 | 1358 | 89 | |
GenBank mRNA | AK298422.1 | - | 5 - 6 | 1179 | 89 |
BC104908.1 | - | 5 - 6 | 1124 | 89 | |
BC104910.1 | - | 5 - 6 | 1124 | 89 | |
BC143483.1 | - | 5 - 6 | 1125 | 89 | |
HM116541.1 | - | 5 - 6 | 1247 | 89 | |
HM116542.1 | - | 5 - 6 | 921 | 89 | |
HM116547.1 | - | 5 - 6 | 1247 | 89 | |
HM116552.1 | - | 6 - 7 | 864 | 89 | |
HM116557.1 | - | 4 - 5 | 1035 | 89 | |
HM116558.1 | - | 4 - 5 | 980 | 89 | |
HM116560.1 | - | 2 - 3 | 99 | 89 | |
HM449088.1 | - | 5 - 6 | 1127 | 89 | |
HM449089.1 | - | 4 - 5 | 927 | 89 | |
M57732.1 | - | 5 - 6 | 1127 | 89 | |
X71346.1 | - | 5 - 6 | 1300 | 89 | |
X71347.1 | - | 5 - 6 | 1235 | 89 |