Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206999.1 | NP_001193928.1 | 7 - 8 | 807 | 84 |
NM_007174.2 | NP_009105.1 | 7 - 8 | 807 | 84 | |
XM_006719206.2 | XP_006719269.1 | 7 - 8 | 834 | 84 | |
XM_011537783.1 | XP_011536085.1 | 7 - 8 | 834 | 84 | |
XM_011537784.1 | XP_011536086.1 | 7 - 8 | 834 | 84 | |
XM_011537785.1 | XP_011536087.1 | 7 - 8 | 834 | 84 | |
XM_011537787.1 | XP_011536089.1 | 7 - 8 | 834 | 84 | |
XM_011537788.1 | XP_011536090.1 | 7 - 8 | 834 | 84 | |
XM_017018735.1 | XP_016874224.1 | 7 - 8 | 834 | 84 | |
XM_017018736.1 | XP_016874225.1 | 7 - 8 | 834 | 84 | |
XM_017018737.1 | XP_016874226.1 | 7 - 8 | 834 | 84 | |
GenBank mRNA | AY209000.1 | - | 7 - 8 | 804 | 84 |
AY257469.1 | - | 7 - 8 | 804 | 84 | |
AY681966.1 | - | 7 - 8 | 807 | 84 |