Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008895.2 | NP_001008895.1 | 14 - 15 | 1659 | 70 |
NM_001278513.1 | NP_001265442.1 | 14 - 15 | 1803 | 70 | |
NM_001278514.1 | NP_001265443.1 | 14 - 15 | 1471 | 70 | |
NM_003589.2 | NP_003580.1 | 14 - 15 | 1485 | 70 | |
XM_011537523.2 | XP_011535825.1 | 13 - 14 | 2019 | 70 | |
GenBank mRNA | AB012193.1 | - | 8 - 9 | 833 | 70 |
AB178950.1 | - | 14 - 15 | 1536 | 70 | |
AF077188.1 | - | 14 - 15 | 1396 | 70 | |
AK296700.1 | - | 6 - 7 | 780 | 70 | |
AL833355.1 | - | 13 - 14 | 1423 | 70 | |
AY365124.1 | - | 14 - 15 | 1536 | 70 | |
BC008308.1 | - | 14 - 15 | 1547 | 70 | |
U58090.1 | - | 4 - 5 | 523 | 70 |