Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000484.3 | NP_000475.1 | 4 - 5 | 671 | 92 |
NM_001136016.3 | NP_001129488.1 | 4 - 5 | 652 | 92 | |
NM_001136129.2 | NP_001129601.1 | 3 - 4 | 503 | 92 | |
NM_001136130.2 | NP_001129602.1 | 3 - 4 | 503 | 92 | |
NM_001136131.2 | NP_001129603.1 | 4 - 5 | 558 | 92 | |
NM_001204301.1 | NP_001191230.1 | 4 - 5 | 671 | 92 | |
NM_001204302.1 | NP_001191231.1 | 4 - 5 | 671 | 92 | |
NM_001204303.1 | NP_001191232.1 | 4 - 5 | 671 | 92 | |
NM_201413.2 | NP_958816.1 | 4 - 5 | 671 | 92 | |
NM_201414.2 | NP_958817.1 | 4 - 5 | 671 | 92 | |
GenBank mRNA | A02759.1 | - | 4 - 5 | 617 | 92 |
AF282245.1 | - | 3 - 4 | 430 | 92 | |
AK294534.1 | - | 3 - 4 | 450 | 92 | |
AK295621.1 | - | 4 - 5 | 613 | 92 | |
AK296229.1 | - | 4 - 5 | 558 | 92 | |
AK298861.1 | - | 3 - 4 | 573 | 92 | |
AK311717.1 | - | 4 - 5 | 731 | 92 | |
AK312326.1 | - | 4 - 5 | 536 | 92 | |
BC004369.1 | - | 4 - 5 | 597 | 92 | |
BC065529.1 | - | 4 - 5 | 602 | 92 | |
M28373.1 | - | 2 - 3 | 234 | 92 | |
X06989.1 | - | 4 - 5 | 595 | 92 | |
Y00264.1 | - | 4 - 5 | 618 | 92 |