Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000484.3 | NP_000475.1 | 7 - 8 | 1236 | 60 |
NM_001136130.2 | NP_001129602.1 | 6 - 7 | 1068 | 60 | |
NM_001204301.1 | NP_001191230.1 | 7 - 8 | 1236 | 60 | |
GenBank mRNA | AK294534.1 | - | 6 - 7 | 1015 | 60 |
AK312326.1 | - | 7 - 8 | 1101 | 60 | |
X06981.1 | - | 1 - 2 | 178 | 60 |