Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282778.1 | NP_001269707.1 | 16 - 17 | 1745 | 68 |
XM_017002004.1 | XP_016857493.1 | 15 - 16 | 1838 | 68 | |
XM_017002008.1 | XP_016857497.1 | 17 - 18 | 1769 | 68 | |
XM_017002009.1 | XP_016857498.1 | 15 - 16 | 1669 | 68 | |
XM_017002010.1 | XP_016857499.1 | 16 - 17 | 1454 | 68 | |
XM_017002013.1 | XP_016857502.1 | 13 - 14 | 2848 | 68 | |
GenBank mRNA | AF090116.1 | - | 16 - 17 | 1600 | 68 |
AK314862.1 | - | 16 - 17 | 1606 | 68 | |
AY587875.1 | - | 15 - 16 | 1363 | 68 | |
U32439.1 | - | 14 - 15 | 1217 | 68 |