Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103.3 | NP_001094.1 | 12 - 13 | 1626 | 84 |
NM_001278343.1 | NP_001265272.1 | 12 - 13 | 1626 | 84 | |
NM_001278344.1 | NP_001265273.1 | 14 - 15 | 1823 | 84 | |
GenBank mRNA | AB209521.1 | - | 14 - 15 | 1767 | 84 |
AK297618.1 | - | 5 - 6 | 708 | 84 | |
AK297628.1 | - | 5 - 6 | 486 | 84 | |
AK315250.1 | - | 12 - 13 | 1626 | 84 | |
BC047901.2 | - | 12 - 13 | 1558 | 84 | |
BC051770.1 | - | 12 - 13 | 1571 | 84 | |
M86406.1 | - | 12 - 13 | 1579 | 84 |