Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_197941.3 | NP_922932.2 | 2 - 3 | 958 | 115 |
NR_135689.1 | 2 - 3 | 958 | 115 | ||
XM_011543113.2 | XP_011541415.1 | 2 - 3 | 973 | 115 | |
XM_011543114.2 | XP_011541416.1 | 2 - 3 | 552 | 115 | |
XM_011543115.2 | XP_011541417.1 | 2 - 3 | 1011 | 115 | |
XM_011543116.2 | XP_011541418.1 | 2 - 3 | 1008 | 115 | |
XM_011543119.2 | XP_011541421.1 | 2 - 3 | 1002 | 115 | |
XM_011543120.2 | XP_011541422.1 | 2 - 3 | 999 | 115 | |
XM_011543122.2 | XP_011541424.1 | 2 - 3 | 999 | 115 | |
XM_011543126.2 | XP_011541428.1 | 2 - 3 | 996 | 115 | |
XM_011543127.2 | XP_011541429.1 | 2 - 3 | 996 | 115 | |
GenBank mRNA | AB209685.1 | - | 2 - 3 | 455 | 115 |
AF140674.1 | - | 1 - 2 | 122 | 115 | |
AY692424.1 | - | 2 - 3 | 916 | 115 | |
AY692426.1 | - | 1 - 2 | 119 | 115 |