Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_197941.3 | NP_922932.2 | 9 - 10 | 2081 | 83 |
NR_135689.1 | 9 - 10 | 2086 | 83 | ||
XM_011543113.2 | XP_011541415.1 | 9 - 10 | 2096 | 83 | |
XM_011543114.2 | XP_011541416.1 | 9 - 10 | 1675 | 83 | |
XM_011543115.2 | XP_011541417.1 | 9 - 10 | 2134 | 83 | |
XM_011543116.2 | XP_011541418.1 | 9 - 10 | 2131 | 83 | |
XM_011543117.1 | XP_011541419.1 | 7 - 8 | 1434 | 83 | |
XM_011543119.2 | XP_011541421.1 | 9 - 10 | 2125 | 83 | |
XM_011543120.2 | XP_011541422.1 | 9 - 10 | 2122 | 83 | |
XM_011543121.2 | XP_011541423.2 | 5 - 6 | 462 | 83 | |
XM_011543122.2 | XP_011541424.1 | 9 - 10 | 2122 | 83 | |
XM_011543123.2 | XP_011541425.1 | 3 - 4 | 200 | 83 | |
XM_011543126.2 | XP_011541428.1 | 9 - 10 | 2119 | 83 | |
XM_011543127.2 | XP_011541429.1 | 9 - 10 | 2119 | 83 | |
GenBank mRNA | AB209685.1 | - | 9 - 10 | 1583 | 83 |
AY692424.1 | - | 9 - 10 | 2039 | 83 | |
AY692425.1 | - | 8 - 9 | 1243 | 83 | |
AY692426.1 | - | 8 - 9 | 1247 | 83 |