Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005481.2 | NP_005472.2 | 7 - 8 | 1291 | 79 |
XM_017026120.1 | XP_016881609.1 | 7 - 8 | 1286 | 79 | |
XM_017026121.1 | XP_016881610.1 | 6 - 7 | 1131 | 79 | |
GenBank mRNA | AF106934.1 | - | 7 - 8 | 1278 | 79 |
AF121228.1 | - | 6 - 7 | 1140 | 79 | |
BC004554.1 | - | 7 - 8 | 1272 | 79 | |
BC007853.2 | - | 3 - 4 | 401 | 79 | |
BC011841.1 | - | 7 - 8 | 1272 | 79 | |
BC017282.2 | - | 7 - 8 | 1266 | 79 | |
EU392449.1 | - | 6 - 7 | 1157 | 79 | |
EU392451.1 | - | 5 - 6 | 719 | 79 |