Assay Design



Gene Transcripts

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
Chromosome Location
Chr.11: 47465937 - 47565569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025596.2 NP_001020767.1 8 - 9 905 63
NM_001172639.1 NP_001166110.1 11 - 12 1867 63
NM_001172640.1 NP_001166111.1 8 - 9 902 63
NM_006560.3 NP_006551.1 9 - 10 1142 63
XM_011519850.2 XP_011518152.2 9 - 10 1216 63
XM_017017105.1 XP_016872594.1 10 - 11 1129 63
XM_017017106.1 XP_016872595.1 12 - 13 1317 63
XM_017017107.1 XP_016872596.1 10 - 11 1129 63
XM_017017111.1 XP_016872600.1 10 - 11 1117 63
XM_017017112.1 XP_016872601.1 10 - 11 1117 63
XM_017017113.1 XP_016872602.1 10 - 11 1117 63
XM_017017114.1 XP_016872603.1 10 - 11 1117 63
XM_017017124.1 XP_016872613.1 11 - 12 1248 63
XM_017017125.1 XP_016872614.1 9 - 10 977 63
XM_017017126.1 XP_016872615.1 9 - 10 1106 63
XM_017017127.1 XP_016872616.1 9 - 10 1130 63
XM_017017128.1 XP_016872617.1 9 - 10 1154 63
XM_017017129.1 XP_016872618.1 10 - 11 1152 63
XM_017017130.1 XP_016872619.1 9 - 10 1106 63
XM_017017131.1 XP_016872620.1 9 - 10 1130 63
XM_017017132.1 XP_016872621.1 9 - 10 1154 63
XM_017017135.1 XP_016872624.1 9 - 10 1094 63
GenBank mRNA AB210019.1 - 11 - 12 1867 63
AF248648.1 - 8 - 9 913 63
AF267533.1 - 8 - 9 906 63
AJ007988.1 - 8 - 9 895 63
AK304430.1 - 8 - 9 845 63
AK307679.1 - 3 - 4 367 63
U63289.1 - 9 - 10 1020 63

View Full Product Details