Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011519848.2 | XP_011518150.1 | 13 - 14 | 1589 | 66 |
XM_017017103.1 | XP_016872592.1 | 14 - 15 | 1741 | 66 | |
XM_017017106.1 | XP_016872595.1 | 16 - 17 | 1926 | 66 | |
XM_017017113.1 | XP_016872602.1 | 14 - 15 | 1726 | 66 | |
XM_017017122.1 | XP_016872611.1 | 13 - 14 | 1718 | 66 |